Learn More
Medchemexpress LLC DMT-2'-O-MOE-rA(Bz) phosphoramidite | 251647-53-7 | MFCD29472587 | 98.0% | 917.98 g·mol⁻¹ | C49H56N7O9P | 250 MG
SDP
Catalog No. 5000243610
Encompass_Preferred
DMT-2'-O-MOE-rA(Bz) phosphoramidite is an adenosine nucleoside phosphoramidite monomer used to introduce 2'-O-methoxyethyl-modified adenosine residues during automated solid-phase oligonucleotide synthesis. It contains a 5'-dimethoxytrityl (DMT) protecting group, an N6-benzoyl base protection, and a 3'-2-cyanoethyl phosphoramidite suitable for standard coupling cycles.
  • Used in solid-phase oligonucleotide synthesis.
  • Incorporates 2'-O-methoxyethyl-modified adenosine to enhance nuclease resistance and binding affinity.
  • 5'-Dimethoxytrityl and N6-benzoyl protections for compatibility with automated synthesis workflows.
  • Available as a solid or as a DMSO solution for convenient handling.
  • Typical purity 98.0% as provided on certificate of analysis.
  • Store at -20 °C under inert atmosphere to maintain stability.
Catalog No. 50-002-43610 Supplier Medchemexpress LLC Supplier No. HY104017250MG
DMT-2'-O-MOE-rA(Bz) phosphoramidite is an adenosine nucleoside phosphoramidite monomer used to introduce 2'-O-methoxyethyl-modified adenosine residues during automated solid-phase oligonucleotide synthesis. It contains a 5'-dimethoxytrityl (DMT) protecting group, an N6-benzoyl base protection, and a 3'-2-cyanoethyl phosphoramidite suitable for standard coupling cycles.
  • Used in solid-phase oligonucleotide synthesis.
  • Incorporates 2'-O-methoxyethyl-modified adenosine to enhance nuclease resistance and binding affinity.
  • 5'-Dimethoxytrityl and N6-benzoyl protections for compatibility with automated synthesis workflows.
  • Available as a solid or as a DMSO solution for convenient handling.
  • Typical purity 98.0% as provided on certificate of analysis.
  • Store at -20 °C under inert atmosphere to maintain stability.