Learn More
Medchemexpress LLC DMT-2'O-MOE-rG(ib) phosphoramidite | 251647-55-9 | MFCD00792625 | >98.0% | 914.01 g/mol | C47H60N7O10P | 250 MG
SDP
Catalog No. 5000241880
Encompass_Preferred
DMT-2'O-MOE-rG(ib) phosphoramidite is a DMT-protected 2'-O-methoxyethoxy (MOE) guanosine phosphoramidite used as a monomer for automated solid-phase oligonucleotide synthesis, including stereochemical synthesis of phosphorothioate oligonucleotides. Supplied for research use, it is offered in solid and solution formats for flexibility in synthesis workflows.
  • DMT-protected 2'-O-methoxyethoxy guanosine phosphoramidite for oligonucleotide assembly.
  • Supports stereochemical synthesis of phosphorothioate oligonucleotides.
  • Available as solid powder or 10 mM solution in DMSO for convenience.
  • High purity suitable for synthesis workflows (reported ≥98% by HPLC).
  • Suitable for incorporation of 2'-O-MOE-modified guanosine residues.
Catalog No. 50-002-41880 Supplier Medchemexpress LLC Supplier No. HY104014250MG
DMT-2'O-MOE-rG(ib) phosphoramidite is a DMT-protected 2'-O-methoxyethoxy (MOE) guanosine phosphoramidite used as a monomer for automated solid-phase oligonucleotide synthesis, including stereochemical synthesis of phosphorothioate oligonucleotides. Supplied for research use, it is offered in solid and solution formats for flexibility in synthesis workflows.
  • DMT-protected 2'-O-methoxyethoxy guanosine phosphoramidite for oligonucleotide assembly.
  • Supports stereochemical synthesis of phosphorothioate oligonucleotides.
  • Available as solid powder or 10 mM solution in DMSO for convenience.
  • High purity suitable for synthesis workflows (reported ≥98% by HPLC).
  • Suitable for incorporation of 2'-O-MOE-modified guanosine residues.