Learn More
Medchemexpress LLC DMT-2 -OMe-Bz-C 1g
SDP
Catalog No. 5000206329
Encompass_Preferred
DMT-2 -OMe-Bz-C is a cytidine analog Cytidine analogs have a mechanism of inhibiting DNA methyltransferases (such as Zebularine HY-13420) and have potential anti-metabolic and anti-tumor activities[1]
Catalog No. 50-002-06329 Supplier Medchemexpress LLC Supplier No. HYW1151861G
DMT-2 -OMe-Bz-C is a cytidine analog Cytidine analogs have a mechanism of inhibiting DNA methyltransferases (such as Zebularine HY-13420) and have potential anti-metabolic and anti-tumor activities[1]