Learn More
Medchemexpress LLC DMT-2'-OMe-D-Ribitol phosphoramidite | 1470023-72-3 | 99.2% | 650.74 | 100 MG
SDP
Catalog No. 5000393153
Encompass_Preferred
DMT-2'-OMe-D-Ribitol phosphoramidite is an abasic phosphoramidite monomer designed for introducing the abasic group Y34 into oligonucleotides. This compound is valuable in molecular biology research, particularly in studies involving RNA duplexes and DNA/RNA synthesis.
  • Abasic phosphoramidite monomer
  • Introduces abasic group Y34 into oligonucleotides
  • Used in DNA/RNA synthesis
  • Relevant to cell cycle/DNA damage research
  • Can be used as an inhibitor in certain applications
Catalog No. 50-003-93153 Supplier Medchemexpress LLC Supplier No. HY160884100MG
DMT-2'-OMe-D-Ribitol phosphoramidite is an abasic phosphoramidite monomer designed for introducing the abasic group Y34 into oligonucleotides. This compound is valuable in molecular biology research, particularly in studies involving RNA duplexes and DNA/RNA synthesis.
  • Abasic phosphoramidite monomer
  • Introduces abasic group Y34 into oligonucleotides
  • Used in DNA/RNA synthesis
  • Relevant to cell cycle/DNA damage research
  • Can be used as an inhibitor in certain applications