Learn More
Medchemexpress LLC DMT-2'-OMe-D-Ribitol phosphoramidite | 1470023-72-3 | 99.2% | 650.74 | 5 MG
SDP
Catalog No. 5000391989
Encompass_Preferred
DMT-2'-OMe-D-Ribitol phosphoramidite (compound 9) is an abasic phosphoramidite monomer that can be used to introduce the abasic group Y34 into oligonucleotides. It is for research use only.
  • An abasic phosphoramidite monomer.
  • Can be used to introduce the abasic group Y34 into oligonucleotides.
  • For research use only.
Catalog No. 50-003-91989 Supplier Medchemexpress LLC Supplier No. HY1608845MG
Add to Cart
Add to Cart
DMT-2'-OMe-D-Ribitol phosphoramidite (compound 9) is an abasic phosphoramidite monomer that can be used to introduce the abasic group Y34 into oligonucleotides. It is for research use only.
  • An abasic phosphoramidite monomer.
  • Can be used to introduce the abasic group Y34 into oligonucleotides.
  • For research use only.