Learn More
Medchemexpress LLC DMT-2'-OMe-D-Ribitol phosphoramidite | 1470023-72-3 | 99.2% | 650.74 | 50 MG
SDP
Catalog No. 5000392620
Encompass_Preferred
DMT-2'-OMe-D-Ribitol phosphoramidite (compound 9) is an abasic phosphoramidite monomer that can be used to introduce the abasic group Y34 into oligonucleotides.
  • Abasic phosphoramidite monomer
  • Introduces abasic group Y34 into oligonucleotides
Catalog No. 50-003-92620 Supplier Medchemexpress LLC Supplier No. HY16088450MG
DMT-2'-OMe-D-Ribitol phosphoramidite (compound 9) is an abasic phosphoramidite monomer that can be used to introduce the abasic group Y34 into oligonucleotides.
  • Abasic phosphoramidite monomer
  • Introduces abasic group Y34 into oligonucleotides