Learn More
Medchemexpress LLC DMT-5Me-dC(Bz)-CE Phosphoramidite | 105931-57-5 | 98.4% | C47H54N5O8P | 1 G
SDP
Catalog No. 5000359201
Encompass_Preferred
DMT-5Me-dC(Bz)-CE Phosphoramidite is a chemical structural unit utilized in solid-phase DNA synthesis. It facilitates the introduction of the 5-methylcytidine base into the DNA chain, thereby enhancing its stability and binding affinity. This product can be employed to construct modified oligonucleotides, specifically for developing locked nucleic acids (LNA).
  • Used in solid-phase DNA synthesis
  • Introduces the 5-methylcytidine base into DNA chains
  • Enhances DNA stability and binding affinity
  • Constructs modified oligonucleotides
  • Useful for developing locked nucleic acids (LNA)
  • Target for DNA/RNA synthesis pathway
  • Related to Cell cycle and DNA damage
Catalog No. 50-003-59201 Supplier Medchemexpress LLC Supplier No. HY463171G
DMT-5Me-dC(Bz)-CE Phosphoramidite is a chemical structural unit utilized in solid-phase DNA synthesis. It facilitates the introduction of the 5-methylcytidine base into the DNA chain, thereby enhancing its stability and binding affinity. This product can be employed to construct modified oligonucleotides, specifically for developing locked nucleic acids (LNA).
  • Used in solid-phase DNA synthesis
  • Introduces the 5-methylcytidine base into DNA chains
  • Enhances DNA stability and binding affinity
  • Constructs modified oligonucleotides
  • Useful for developing locked nucleic acids (LNA)
  • Target for DNA/RNA synthesis pathway
  • Related to Cell cycle and DNA damage