Learn More
Medchemexpress LLC DMT-5Me-dC(Bz)-CE Phosphoramidite | 105931-57-5 | 98.4% | C47H54N5O8P | 1 ML
SDP
Catalog No. 5000357754
Encompass_Preferred
DMT-5Me-dC(Bz)-CE Phosphoramidite is a chemical structural unit designed for solid-phase DNA synthesis. It introduces the 5-methylcytidine base into the DNA chain, enhancing its stability and binding affinity. This compound is suitable for constructing modified oligonucleotides, especially for locked nucleic acids (LNA). It is supplied as a 10 mM solution in DMSO.
  • Chemical structural unit for solid-phase DNA synthesis
  • Introduces 5-methylcytidine base into DNA chain
  • Enhances DNA stability and binding affinity
  • Suitable for constructing modified oligonucleotides
  • Can be used for locked nucleic acids (LNA)
Catalog No. 50-003-57754 Supplier Medchemexpress LLC Supplier No. HY4631710MM/1ML
DMT-5Me-dC(Bz)-CE Phosphoramidite is a chemical structural unit designed for solid-phase DNA synthesis. It introduces the 5-methylcytidine base into the DNA chain, enhancing its stability and binding affinity. This compound is suitable for constructing modified oligonucleotides, especially for locked nucleic acids (LNA). It is supplied as a 10 mM solution in DMSO.
  • Chemical structural unit for solid-phase DNA synthesis
  • Introduces 5-methylcytidine base into DNA chain
  • Enhances DNA stability and binding affinity
  • Suitable for constructing modified oligonucleotides
  • Can be used for locked nucleic acids (LNA)