Learn More
Medchemexpress LLC DMT-5Me-dC(Bz)-CE Phosphoramidite | 105931-57-5 | 98.4% | C47H54N5O8P | 100 MG
SDP
Catalog No. 5000357874
Encompass_Preferred
DMT-5Me-dC(Bz)-CE Phosphoramidite is a chemical structural unit utilized in solid-phase DNA synthesis. It can introduce the 5-methylcytidine base into the DNA chain, thereby enhancing its stability and binding affinity. This compound is suitable for constructing modified oligonucleotides, particularly for the creation of locked nucleic acids (LNA). It is intended for research use only.
  • Utilized in solid-phase DNA synthesis
  • Introduces the 5-methylcytidine base into the DNA chain
  • Enhances DNA stability and binding affinity
  • Suitable for constructing modified oligonucleotides
  • Particularly for the creation of locked nucleic acids (LNA)
  • Intended for research use only
Catalog No. 50-003-57874 Supplier Medchemexpress LLC Supplier No. HY46317100MG
DMT-5Me-dC(Bz)-CE Phosphoramidite is a chemical structural unit utilized in solid-phase DNA synthesis. It can introduce the 5-methylcytidine base into the DNA chain, thereby enhancing its stability and binding affinity. This compound is suitable for constructing modified oligonucleotides, particularly for the creation of locked nucleic acids (LNA). It is intended for research use only.
  • Utilized in solid-phase DNA synthesis
  • Introduces the 5-methylcytidine base into the DNA chain
  • Enhances DNA stability and binding affinity
  • Suitable for constructing modified oligonucleotides
  • Particularly for the creation of locked nucleic acids (LNA)
  • Intended for research use only