Learn More
Medchemexpress LLC DMT-5Me-dC(Bz)-CE Phosphoramidite | 105931-57-5 | 98.4% | C47H54N5O8P | 500 MG
SDP
Catalog No. 5000357755
Encompass_Preferred
DMT-5Me-dC(Bz)-CE Phosphoramidite is a chemical structural unit utilized in solid-phase DNA synthesis. It introduces the 5-methylcytidine base into the DNA chain to enhance its stability and binding affinity. This compound is suitable for constructing modified oligonucleotides, particularly for creating locked nucleic acids (LNA).
  • Facilitates the introduction of the 5-methylcytidine base into the DNA chain
  • Enhances DNA chain stability and binding affinity
  • Suitable for constructing modified oligonucleotides
  • Useful for creating locked nucleic acids (LNA)
  • Intended for research use only
Catalog No. 50-003-57755 Supplier Medchemexpress LLC Supplier No. HY46317500MG
DMT-5Me-dC(Bz)-CE Phosphoramidite is a chemical structural unit utilized in solid-phase DNA synthesis. It introduces the 5-methylcytidine base into the DNA chain to enhance its stability and binding affinity. This compound is suitable for constructing modified oligonucleotides, particularly for creating locked nucleic acids (LNA).
  • Facilitates the introduction of the 5-methylcytidine base into the DNA chain
  • Enhances DNA chain stability and binding affinity
  • Suitable for constructing modified oligonucleotides
  • Useful for creating locked nucleic acids (LNA)
  • Intended for research use only