Learn More
Medchemexpress LLC DMT-dA(PAc) Phosphoramidite | 110543-74-3 | 98.8% | 887.96 | 100 MG
SDP
Catalog No. 5000350415
Encompass_Preferred
This compound is a phosphoramidite, a synthetic building block utilized in the chemical synthesis of DNA or RNA. It serves as a precursor in oligonucleotide synthesis, enabling the precise construction of nucleic acid chains. The substance has a molecular weight of 887.96 and a chemical formula of C48H54N7O8P. It typically appears as a white to off-white solid. It is soluble in DMSO at 100 mg/mL.
  • Chemical purity: 98.75%, ensuring reliable results.
  • Application in nucleic acid synthesis for molecular biology research and therapeutic development.
  • Versatile solubility: it can be prepared as a solution in DMSO, with protocols for suspended and clear solutions for in vivo studies.
  • Detailed chemical characterization for accurate identification and use.
Catalog No. 50-003-50415 Supplier Medchemexpress LLC Supplier No. HY138581100MG
This compound is a phosphoramidite, a synthetic building block utilized in the chemical synthesis of DNA or RNA. It serves as a precursor in oligonucleotide synthesis, enabling the precise construction of nucleic acid chains. The substance has a molecular weight of 887.96 and a chemical formula of C48H54N7O8P. It typically appears as a white to off-white solid. It is soluble in DMSO at 100 mg/mL.
  • Chemical purity: 98.75%, ensuring reliable results.
  • Application in nucleic acid synthesis for molecular biology research and therapeutic development.
  • Versatile solubility: it can be prepared as a solution in DMSO, with protocols for suspended and clear solutions for in vivo studies.
  • Detailed chemical characterization for accurate identification and use.