Learn More
Medchemexpress LLC DMT-dC(ac) Phosphoramidite | 154110-40-4 | 99.76% | 771.84 | 1 G
SDP
Catalog No. 5000351476
Encompass_Preferred
This phosphoramidite derivative is utilized in oligonucleotide synthesis for research purposes. It comes as a white to off-white solid with a purity of 99.76%. For storage, it should be kept at -20°C, protected from light, and under nitrogen. When dissolved, stock solutions can be stored at -80°C for up to 6 months or -20°C for 1 month, also protected from light and under nitrogen.
  • Derivative for creating oligonucleotides
  • Suitable for research applications
  • High purity for reliable experimental results
  • Can be dissolved in DMSO, and various in vivo protocols are available using co-solvents like PEG300, Tween-80, Saline, or Corn Oil
Catalog No. 50-003-51476 Supplier Medchemexpress LLC Supplier No. HY1385861G
This phosphoramidite derivative is utilized in oligonucleotide synthesis for research purposes. It comes as a white to off-white solid with a purity of 99.76%. For storage, it should be kept at -20°C, protected from light, and under nitrogen. When dissolved, stock solutions can be stored at -80°C for up to 6 months or -20°C for 1 month, also protected from light and under nitrogen.
  • Derivative for creating oligonucleotides
  • Suitable for research applications
  • High purity for reliable experimental results
  • Can be dissolved in DMSO, and various in vivo protocols are available using co-solvents like PEG300, Tween-80, Saline, or Corn Oil