Learn More
Medchemexpress LLC DMT-dC(ac) Phosphoramidite | 154110-40-4 | 99.8% | 771.84 | 10 G
SDP
Catalog No. 5000350019
Encompass_Preferred
DMT-dC(ac) Phosphoramidite (compound 30f) is a phosphoramidite derivative that can be used for oligonucleotide synthesis. It is intended for research use only.
  • Can be used for oligonucleotide synthesis.
  • Intended for research use only.
Catalog No. 50-003-50019 Supplier Medchemexpress LLC Supplier No. HY13858610G
DMT-dC(ac) Phosphoramidite (compound 30f) is a phosphoramidite derivative that can be used for oligonucleotide synthesis. It is intended for research use only.
  • Can be used for oligonucleotide synthesis.
  • Intended for research use only.