Learn More
Sigma Aldrich Fine Chemicals Biosciences DMT-dC(ac) Phosphoramidite configured for ABI | 154110-40-4 | MFCD09842112 | 6X5G
Catalog No. 501744076
Encompass
DMT-dC(ac) Phosphoramidite configured for ABI | Purity: >=98% (31P-NMR); >=99.0% (reversed phase HPLC) | Mol Wt: 771.84 | 154110-40-4 | MFCD09842112 | 6X5G
Catalog No. 50-174-4076 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C1130506X5G
DMT-dC(ac) Phosphoramidite configured for ABI | Purity: >=98% (31P-NMR); >=99.0% (reversed phase HPLC) | Mol Wt: 771.84 | 154110-40-4 | MFCD09842112 | 6X5G