Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DMT-dC(bz) Phosphoramidite | 102212-98-6 | MFCD00036315 | 99.4% | 833.91 | 1 G
SDP

Catalog No. 5000453672
Encompass_Preferred

DMT-dC(bz) Phosphoramidite is a deoxycytidine phosphoramide monomer with a protective group. It is commonly used for synthesizing DNA dimers or trimers.

  • Appearance: Solid
  • Color: White to off-white
  • Formula: C46H52N5O8P

Catalog No. 50-004-53672 Supplier Medchemexpress LLC Supplier No. HYW0088491G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DMT-dC(bz) Phosphoramidite is a deoxycytidine phosphoramide monomer with a protective group. It is commonly used for synthesizing DNA dimers or trimers.

  • Appearance: Solid
  • Color: White to off-white
  • Formula: C46H52N5O8P

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.