Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DMT-dG(dmf) Phosphoramidite | 330628-04-1 | 98.8% | 824.90 | 10 G
SDP

Catalog No. 5000352452
Encompass_Preferred

DMT-dG(dmf) Phosphoramidite is a phosphinamide monomer that plays a crucial role in the preparation of oligonucleotides. This compound is strictly for research purposes and not intended for patient use.

  • Phosphinamide monomer
  • Supports oligonucleotide synthesis
  • For research use only
  • Targets DNA/RNA synthesis pathways
  • Involved in cell cycle and DNA damage pathways
  • Store at -20°C, protected from light
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C (protect from light)
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-52452 Supplier Medchemexpress LLC Supplier No. HY13858510G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DMT-dG(dmf) Phosphoramidite is a phosphinamide monomer that plays a crucial role in the preparation of oligonucleotides. This compound is strictly for research purposes and not intended for patient use.

  • Phosphinamide monomer
  • Supports oligonucleotide synthesis
  • For research use only
  • Targets DNA/RNA synthesis pathways
  • Involved in cell cycle and DNA damage pathways
  • Store at -20°C, protected from light
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C (protect from light)
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.