Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DMT-dG(dmf) Phosphoramidite | 330628-04-1 | 98.8% | 824.90 | 25 G
SDP

Catalog No. 5000347837
Encompass_Preferred

DMT-dG(dmf) Phosphoramidite is a phosphinamide monomer that can be used in the preparation of oligonucleotides. It is intended for research use only.

  • Purity of 98.8%
  • Molecular weight of 824.90
  • Chemical formula C43H53N8O7P
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL
  • Utilized in the preparation of oligonucleotides

Catalog No. 50-003-47837 Supplier Medchemexpress LLC Supplier No. 25GHY138585
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DMT-dG(dmf) Phosphoramidite is a phosphinamide monomer that can be used in the preparation of oligonucleotides. It is intended for research use only.

  • Purity of 98.8%
  • Molecular weight of 824.90
  • Chemical formula C43H53N8O7P
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL
  • Utilized in the preparation of oligonucleotides

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.