Learn More
Medchemexpress LLC DMT-dG(ib) Phosphoramidite | 93183-15-4 | 99.7% | 839.92 | 10 G
SDP
Catalog No. 5000451201
Encompass_Preferred
DMT-dG(ib) Phosphoramidite is a phosphoramidite monomer utilized in the synthesis of DNA and related molecules. This product is for research use only.
  • Monomer for synthesis of DNA
  • Monomer for synthesis of related molecules
  • For research use only
Catalog No. 50-004-51201 Supplier Medchemexpress LLC Supplier No. HYW00884810G
DMT-dG(ib) Phosphoramidite is a phosphoramidite monomer utilized in the synthesis of DNA and related molecules. This product is for research use only.
  • Monomer for synthesis of DNA
  • Monomer for synthesis of related molecules
  • For research use only