Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DMT-dI Phosphoramidite | 141684-35-7 | >98.1% | 754.81 | 1 ML
SDP

Catalog No. 5000349196
Encompass_Preferred

DMT-dI Phosphoramidite is a modified phosphoramidite monomer specifically utilized for oligonucleotide synthesis. It is presented as a solid, appearing in a white to off-white color. The product should be stored at 4°C in a sealed container, away from moisture and light. If stored in solvent, it maintains stability at -80°C for 6 months or at -20°C for 1 month under sealed conditions, also away from moisture and light.

  • Appearance: Solid
  • Color: White to off-white
  • Storage:
    • 4°C, sealed storage, away from moisture and light.
    • In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light).
  • Solubility: Soluble in DMSO at 125 mg/mL (165.60 mM), requiring ultrasonication.

Catalog No. 50-003-49196 Supplier Medchemexpress LLC Supplier No. HY13757610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DMT-dI Phosphoramidite is a modified phosphoramidite monomer specifically utilized for oligonucleotide synthesis. It is presented as a solid, appearing in a white to off-white color. The product should be stored at 4°C in a sealed container, away from moisture and light. If stored in solvent, it maintains stability at -80°C for 6 months or at -20°C for 1 month under sealed conditions, also away from moisture and light.

  • Appearance: Solid
  • Color: White to off-white
  • Storage:
    • 4°C, sealed storage, away from moisture and light.
    • In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light).
  • Solubility: Soluble in DMSO at 125 mg/mL (165.60 mM), requiring ultrasonication.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.