Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DMT-dI Phosphoramidite | 141684-35-7 | 98.15% | 754.81 | 100 MG
SDP

Catalog No. 5000352263
Encompass_Preferred

DMT-dI Phosphoramidite is a modified phosphoramidite monomer designed for oligonucleotide synthesis. This product is for research use only.

  • Modified phosphoramidite monomer
  • Used for oligonucleotide synthesis
  • Purity of 98.15%
  • Molecular weight of 754.81
  • Appears as a white to off-white solid

Catalog No. 50-003-52263 Supplier Medchemexpress LLC Supplier No. HY137576100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DMT-dI Phosphoramidite is a modified phosphoramidite monomer designed for oligonucleotide synthesis. This product is for research use only.

  • Modified phosphoramidite monomer
  • Used for oligonucleotide synthesis
  • Purity of 98.15%
  • Molecular weight of 754.81
  • Appears as a white to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.