Learn More
Medchemexpress LLC DMT-dT Phosphoramidite | 98796-51-1 | 98.8% | 744.81 | 5 G
SDP
Catalog No. 5000454614
Encompass_Preferred
DMT-dT Phosphoramidite is typically used in the synthesis of DNA. It acts as a protecting group and synthetic intermediate in DNA synthesis. The Dimethoxytrityl (DMT) group protects the hydroxyl (-OH) group during DNA synthesis to prevent unwanted reactions. The phosphoramidite interacts with the hydroxyl group (-OH) at the 5' end of the DNA chain to form a new phosphodiester bond, thereby extending the DNA chain.
  • Used for DNA synthesis.
  • Functions as a protecting group.
  • Serves as a synthetic intermediate.
  • Dimethoxytrityl (DMT) group protects hydroxyl group.
  • Extends DNA chain by forming new phosphodiester bonds.
  • Can be used for surface functionalization.
  • Can be used for DNA probe synthesis.
Catalog No. 50-004-54614 Supplier Medchemexpress LLC Supplier No. HYW0130685G
DMT-dT Phosphoramidite is typically used in the synthesis of DNA. It acts as a protecting group and synthetic intermediate in DNA synthesis. The Dimethoxytrityl (DMT) group protects the hydroxyl (-OH) group during DNA synthesis to prevent unwanted reactions. The phosphoramidite interacts with the hydroxyl group (-OH) at the 5' end of the DNA chain to form a new phosphodiester bond, thereby extending the DNA chain.
  • Used for DNA synthesis.
  • Functions as a protecting group.
  • Serves as a synthetic intermediate.
  • Dimethoxytrityl (DMT) group protects hydroxyl group.
  • Extends DNA chain by forming new phosphodiester bonds.
  • Can be used for surface functionalization.
  • Can be used for DNA probe synthesis.