Learn More
Kyfora Bio DMTAP 25 MG
Catalog No. 502559833
Encompass_Preferred
DMTAP Chloride (1,2-dimyristoyl-3-trimethylammonium-propane chloride) is a cationic lipid and an analogue of DOTAP. It can be used to formulate lipid nanoparticles (LNPs) and lipid nanodiscs for the delivery nucleic acids, such as siRNA as well as for magnetic cationic liposomes for targeting tumor microvasculature. Key application(s): Drug delivery, nucleic acid delivery
Catalog No. 50-255-9833 Supplier Kyfora Bio Supplier No. 1448825
DMTAP Chloride (1,2-dimyristoyl-3-trimethylammonium-propane chloride) is a cationic lipid and an analogue of DOTAP. It can be used to formulate lipid nanoparticles (LNPs) and lipid nanodiscs for the delivery nucleic acids, such as siRNA as well as for magnetic cationic liposomes for targeting tumor microvasculature. Key application(s): Drug delivery, nucleic acid delivery