Learn More
Aviva Systems Biology Corp DMTF1 Antibody - C-terminal region (ARP34602_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34602_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DMTF1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human DMTF1. Alternate names: DMP1, DMTF, FLJ41265, hDMP1ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55475