Learn More
Aviva Systems Biology Corp DMTF1 Antibody - N-terminal region (ARP39485_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39485_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DMTF1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human DMTF1. Alternate names: DMP1, DMTF, FLJ25188, FLJ41265, FLJ76054, hDMP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-812