Learn More
Aviva Systems Biology Corp DMTF1 Antibody - middle region (ARP34828_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34828_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DMTF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DMTF1. Alternate names: DMP1, DMTF, FLJ41265, hDMP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55578