Learn More
FUJIFILM Biosciences DMTP Oxon Standard
Catalog No. 5000450357
Encompass_Preferred
High-purity DMTP Oxon standard suitable for pesticide residue analysis Validated by GC and qNMR with 98% assay Packaged under inert gas and stored at 2 10 C
Catalog No. 50-004-50357 Supplier FUJIFILM Biosciences Supplier No. 04930381
High-purity DMTP Oxon standard suitable for pesticide residue analysis Validated by GC and qNMR with 98% assay Packaged under inert gas and stored at 2 10 C