Learn More
Medchemexpress LLC DMTr-LNA-5MeU-3-CED-phosphoramidite | 206055-75-6 | 98.8% | 772.82 | 250 MG
SDP
Catalog No. 5000347751
Encompass_Preferred
DMTr-LNA-5MeU-3-CED-phosphoramidite is a nucleoside derivative. It is for research use only, suitable for various laboratory applications. Key features include:
  • Designed for solid-phase oligonucleotide synthesis
  • Contains 5'-O-4,4'-dimethoxytrityl protecting group
  • Incorporates 5-methyluracil nucleobase
  • Features locked nucleic acid (LNA) modification
Catalog No. 50-003-47751 Supplier Medchemexpress LLC Supplier No. 250MGHY111531
DMTr-LNA-5MeU-3-CED-phosphoramidite is a nucleoside derivative. It is for research use only, suitable for various laboratory applications. Key features include:
  • Designed for solid-phase oligonucleotide synthesis
  • Contains 5'-O-4,4'-dimethoxytrityl protecting group
  • Incorporates 5-methyluracil nucleobase
  • Features locked nucleic acid (LNA) modification