Learn More
Medchemexpress LLC DMTr-TNA-G(O6-CONPh2)(N2Ac)-amidite | 325683-97-4 | 98.2% | 993.05 | C54H57N8O9P | 10 MG
SDP
Catalog No. 5000285854
Encompass_Preferred
Protected TNA guanine amidite for solid-phase oligonucleotide synthesis. This purine nucleoside analog is designed for incorporation into synthetic oligonucleotides and is used in research on nucleic acid chemistry and therapeutic oligonucleotides. The compound is supplied at research-scale quantities and exhibits high chemical purity.
  • Used in solid-phase oligonucleotide synthesis
  • Threose nucleic acid (TNA) guanine building block
  • Protected amidite enabling standard phosphoramidite coupling chemistry
  • High purity: 98.2%
  • Molecular weight: 993.05
  • Suitable for research into nucleic acid analogs and antitumor studies
Catalog No. 50-002-85854 Supplier Medchemexpress LLC Supplier No. HY15459210MG
Protected TNA guanine amidite for solid-phase oligonucleotide synthesis. This purine nucleoside analog is designed for incorporation into synthetic oligonucleotides and is used in research on nucleic acid chemistry and therapeutic oligonucleotides. The compound is supplied at research-scale quantities and exhibits high chemical purity.
  • Used in solid-phase oligonucleotide synthesis
  • Threose nucleic acid (TNA) guanine building block
  • Protected amidite enabling standard phosphoramidite coupling chemistry
  • High purity: 98.2%
  • Molecular weight: 993.05
  • Suitable for research into nucleic acid analogs and antitumor studies