Learn More
Medchemexpress LLC HY-101284 100mg Medchemexpress, DMU2105 CAS:1821143-79-6 Purity:>98%
SDP
Catalog No. 501932088
Encompass_Preferred
Medchemexpress, HY-101284 100mg DMU2105 CAS:1821143-79-6 DMU2105 is a potent and specific CYP1B1 inhibitor, with IC 50 s of 10 nM and 742 nM for CYP1B1 and CYP1A1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2088 Supplier Medchemexpress LLC Supplier No. HY101284100MG
Medchemexpress, HY-101284 100mg DMU2105 CAS:1821143-79-6 DMU2105 is a potent and specific CYP1B1 inhibitor, with IC 50 s of 10 nM and 742 nM for CYP1B1 and CYP1A1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.