Learn More
Medchemexpress LLC HY-101285 100mg Medchemexpress, DMU2139 CAS:1821143-80-9 Purity:>98%
SDP
Catalog No. 501932089
Encompass_Preferred
Medchemexpress, HY-101285 100mg DMU2139 CAS:1821143-80-9 DMU2139 is a potent and specific CYP1B1 inhibitor, with IC 50 s of 9 nM and 795 nM for CYP1B1 and CYP1A1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2089 Supplier Medchemexpress LLC Supplier No. HY101285100MG
Add to Cart
Add to Cart
Medchemexpress, HY-101285 100mg DMU2139 CAS:1821143-80-9 DMU2139 is a potent and specific CYP1B1 inhibitor, with IC 50 s of 9 nM and 795 nM for CYP1B1 and CYP1A1, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.