Learn More
Medchemexpress LLC HY-111754 5mg Medchemexpress, DMX-5804 CAS:2306178-56-1 Purity:>98%
SDP
Catalog No. 501932389
Encompass_Preferred
Medchemexpress, HY-111754 5mg DMX-5804 CAS:2306178-56-1 DMX-5804 is a potent, orally active and selective MAP4K4 inhibitor, with an IC 50 of 3 nM, a pIC 50 of 8.55 for human MAP4K4, less potent on MINK1/MAP4K6 ( pIC 50 , 8.18), and TNIK/MAP4K7 ( pIC 50 , 7.96). DMX-5804 enhances cardiomyocyte survival, and reduces ischemia-reperfusion injury in mice [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2389 Supplier Medchemexpress LLC Supplier No. HY1117545MG
Medchemexpress, HY-111754 5mg DMX-5804 CAS:2306178-56-1 DMX-5804 is a potent, orally active and selective MAP4K4 inhibitor, with an IC 50 of 3 nM, a pIC 50 of 8.55 for human MAP4K4, less potent on MINK1/MAP4K6 ( pIC 50 , 8.18), and TNIK/MAP4K7 ( pIC 50 , 7.96). DMX-5804 enhances cardiomyocyte survival, and reduces ischemia-reperfusion injury in mice [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.