Learn More
Medchemexpress LLC HY-10964 100mg , DMXAA ASA-404;Vadimezan CAS:117570-53-3 Purity:98%
SDP
Catalog No. 501492949
Encompass_Preferred
Medchemexpress, HY-10964 100mg DMXAA ASA-404;Vadimezan CAS:117570-53-3 Formula:C17H14O4 STING , type I IFNs Purity:98% DMXAA, the vascular disrupting agent, is a murine agonist of the stimulator of interferon genes ( STING ) and also a potent inducer of type I IFNs and other cytokines. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2949 Supplier Medchemexpress LLC Supplier No. HY10964100MG
Medchemexpress, HY-10964 100mg DMXAA ASA-404;Vadimezan CAS:117570-53-3 Formula:C17H14O4 STING , type I IFNs Purity:98% DMXAA, the vascular disrupting agent, is a murine agonist of the stimulator of interferon genes ( STING ) and also a potent inducer of type I IFNs and other cytokines. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.