Learn More
Apexbio Technology LLC DMXAA (Vadimezan) 117570-53-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061583
Encompass_Preferred
DMXAA (Vadimezan AS-1404) is a tumor vascular disrupting and apoptosis-inducing agent functioning primarily through inhibition of DT-diaphorase (DTD) It acts by suppressing DTD enzymatic activity showing an IC50 of approximately 62 5 M Additionally DMXAA inhibits various kinases notably VEGFR2 disrupting endothelial cells and blood vessel formation to induce tumor cell apoptosis and vascular necrosis Experimentally DMXAA demonstrates activity in murine tumor models including induction of apoptosis in endothelial cells and tumor vasculature promoting necrosis and tumor regression Therefore DMXAA is commonly used in oncology research focused on anti-angiogenesis and tumor vascular disruption mechanisms
Catalog No. 50-000-61583 Supplier Apexbio Technology LLC Supplier No. A82335.1
 Restricted Item
Add to Cart
Add to Cart
DMXAA (Vadimezan AS-1404) is a tumor vascular disrupting and apoptosis-inducing agent functioning primarily through inhibition of DT-diaphorase (DTD) It acts by suppressing DTD enzymatic activity showing an IC50 of approximately 62 5 M Additionally DMXAA inhibits various kinases notably VEGFR2 disrupting endothelial cells and blood vessel formation to induce tumor cell apoptosis and vascular necrosis Experimentally DMXAA demonstrates activity in murine tumor models including induction of apoptosis in endothelial cells and tumor vasculature promoting necrosis and tumor regression Therefore DMXAA is commonly used in oncology research focused on anti-angiogenesis and tumor vascular disruption mechanisms