Learn More
Cayman Chemical DmyoInosItol phsphat Scre 50uL
SDP
Catalog No. 502892841
Encompass_Preferred
An inhibitor of wild-type and mutant EGFR (IC50s 1.07 0.46 0.13 0.26 and 0.19 nM for wild-type EGFR EGFRd746-750/T790M/C797S EGFRL858R/T790M/C797S EGFRd746-750/T790M and EGFRL858R/T790M respectively) inhibits the proliferation of wild-type EGFR-expressing A431 EGFRd746-750/T790M/C797S-expressing Ba/F3 EGFRd746-750/T790M/C797S-expressing H1975 and EGFRd746-750-expressing PC-9 cells (IC50s 147 26.8 163 and 45 nM respectively) reduces tumor growth in triple mutant EGFR mouse xenograft models
Catalog No. 50-289-2841 Supplier Cayman Chemical Supplier No. 900389550 UL
An inhibitor of wild-type and mutant EGFR (IC50s 1.07 0.46 0.13 0.26 and 0.19 nM for wild-type EGFR EGFRd746-750/T790M/C797S EGFRL858R/T790M/C797S EGFRd746-750/T790M and EGFRL858R/T790M respectively) inhibits the proliferation of wild-type EGFR-expressing A431 EGFRd746-750/T790M/C797S-expressing Ba/F3 EGFRd746-750/T790M/C797S-expressing H1975 and EGFRd746-750-expressing PC-9 cells (IC50s 147 26.8 163 and 45 nM respectively) reduces tumor growth in triple mutant EGFR mouse xenograft models