Learn More
Cayman Chemical DmyoInositol1 3 4 6tetraphosp
SDP
Catalog No. 502888639
Encompass_Preferred
Largely acts a cell signaling intermediate; serving as substrate to produce IP5s, which can be further phosphorylated to produce Ins(1,2,3,4,5,6-)P6; also phosphorylated to produce phytic acid, which serves diverse roles in eukaryotic tissues; a poor activator of the Ins(1,4,5)-P3 receptor in vitro.
Catalog No. 50-288-8639 Supplier Cayman Chemical Supplier No. 10008442500 UG
Add to Cart
Add to Cart
Largely acts a cell signaling intermediate; serving as substrate to produce IP5s, which can be further phosphorylated to produce Ins(1,2,3,4,5,6-)P6; also phosphorylated to produce phytic acid, which serves diverse roles in eukaryotic tissues; a poor activator of the Ins(1,4,5)-P3 receptor in vitro.