Learn More
Cayman Chemical D-myo-Inositol-1-phosphate (sodium salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502889103
Encompass_Preferred
A synthetic, pure phosphoinositol for signal transduction research; can be formed by PLC hydrolysis of phosphatidylinositol or by dephosphorylation of polyphosphate inositols such as Ins(1,3)P2 by inositol polyphosphate 3-phosphatase
Catalog No. 50-288-9103 Supplier Cayman Chemical Supplier No. 1000777710 MG
A synthetic, pure phosphoinositol for signal transduction research; can be formed by PLC hydrolysis of phosphatidylinositol or by dephosphorylation of polyphosphate inositols such as Ins(1,3)P2 by inositol polyphosphate 3-phosphatase