Learn More
Cayman Chemical DmyoInositol1phosphate sodiu
SDP
Supplier:  Cayman Chemical 10007777500 UG
Encompass_Preferred
A synthetic, pure phosphoinositol for signal transduction research; can be formed by PLC hydrolysis of phosphatidylinositol or by dephosphorylation of polyphosphate inositols such as Ins(1,3)P2 by inositol polyphosphate 3-phosphatase
Catalog No. 50-289-0494