Learn More
Cayman Chemical D-myo-Inositol-1-phosphate (sodium salt), Purity: ≥98%, 1 mg
SDP
Catalog No. 502890483
Encompass_Preferred
A synthetic, pure phosphoinositol for signal transduction research; can be formed by PLC hydrolysis of phosphatidylinositol or by dephosphorylation of polyphosphate inositols such as Ins(1,3)P2 by inositol polyphosphate 3-phosphatase
Catalog No. 50-289-0483 Supplier Cayman Chemical Supplier No. 100077771 MG
A synthetic, pure phosphoinositol for signal transduction research; can be formed by PLC hydrolysis of phosphatidylinositol or by dephosphorylation of polyphosphate inositols such as Ins(1,3)P2 by inositol polyphosphate 3-phosphatase