Learn More
Cayman Chemical DmyoInositol3phosphate sodiu
Supplier: Cayman Chemical 10007778100 UG
Synthetic, pure phosphoinositol for signal transduction research; can be formed by the dephosphorylation of polyphosphate inositols such as Ins(3,4)P2 by inositol polyphosphate 4-phosphatase