Learn More
Cayman Chemical D-myo-Inositol-3-phosphate (sodium salt), Purity: ≥98%, 1 mg
SDP
Catalog No. 502888212
Encompass_Preferred
Synthetic, pure phosphoinositol for signal transduction research; can be formed by the dephosphorylation of polyphosphate inositols such as Ins(3,4)P2 by inositol polyphosphate 4-phosphatase
Catalog No. 50-288-8212 Supplier Cayman Chemical Supplier No. 100077781 MG
Synthetic, pure phosphoinositol for signal transduction research; can be formed by the dephosphorylation of polyphosphate inositols such as Ins(3,4)P2 by inositol polyphosphate 4-phosphatase