Learn More
Cayman Chemical DmyoInositol3phosphate sodiu
SDP
Supplier:  Cayman Chemical 10007778500 UG
Encompass_Preferred
Synthetic, pure phosphoinositol for signal transduction research; can be formed by the dephosphorylation of polyphosphate inositols such as Ins(3,4)P2 by inositol polyphosphate 4-phosphatase
Catalog No. 50-288-7854