Learn More
Cayman Chemical DAN-1 EE (hydrochloride), Purity: ≥95%, CAS: 1049720-51-5, 50 mg
SDP
Catalog No. 502896518
Encompass_Preferred
DAN-1 EE is a fluorescent indicator for the bioimaging of NO.{6534} DAN-1 EE is a cell permeable derivative of DAN, a molecule which has been used extensively in the quantitation of nitrate and nitrite using fluorescence spectroscopy.{2762,1701} Upon entry into the cell, DAN-1 EE is transformed into the less cell permeable DAN-1 by cellular esterases thus preventing loss of signal due to diffusion of the molecule from the cell. Intracellular formation of NO can be monitored using excitation and emission wavelengths of 360-380 nm and 420-450 nm, respectively.{6534}
Catalog No. 50-289-6518 Supplier Cayman Chemical Supplier No. 8507050 MG
DAN-1 EE is a fluorescent indicator for the bioimaging of NO.{6534} DAN-1 EE is a cell permeable derivative of DAN, a molecule which has been used extensively in the quantitation of nitrate and nitrite using fluorescence spectroscopy.{2762,1701} Upon entry into the cell, DAN-1 EE is transformed into the less cell permeable DAN-1 by cellular esterases thus preventing loss of signal due to diffusion of the molecule from the cell. Intracellular formation of NO can be monitored using excitation and emission wavelengths of 360-380 nm and 420-450 nm, respectively.{6534}