Learn More
Molecular Research Center DN127 DNAZOL 100ML
SDP
Catalog No. NC9400601
Encompass
DNAzol,100 ml,Genomic DNA isolation reagent for use with tissues,cells and liquids. Non-toxic,no phenol use,the isolation is fast (10-30 min),simple and reliable. 1 ml of DNAzol isolates DNA from 25-50 mg tissue or 1-3 x 107 cells
Catalog No. NC9400601 Supplier Molecular Research Center Supplier No. DN 127
DNAzol,100 ml,Genomic DNA isolation reagent for use with tissues,cells and liquids. Non-toxic,no phenol use,the isolation is fast (10-30 min),simple and reliable. 1 ml of DNAzol isolates DNA from 25-50 mg tissue or 1-3 x 107 cells