Learn More
Ace Glass, Inc. DN25 Ball joint to 24/40 inner joint adapter
SDP
Catalog No. 50978276
Encompass_Preferred
DN25 Ball joint to 24/40 inner joint adapter
Catalog No. 50-978-276 Supplier Ace Glass, Inc. Supplier No. 502045
DN25 Ball joint to 24/40 inner joint adapter