Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical DNA α-pyrene amidite | 200 mg | CAS 181227-52-1 | InChIKey YYLVWADLYIVQFL-DGAGUGGYSA-N

Catalog No. 502355499
Encompass

DNA α-pyrene amidite is a Amidite reagent (Subcategory: Specialty DNA Amidite) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 181227-52-1
- MDL: No data
- InChIKey: YYLVWADLYIVQFL-DGAGUGGYSA-N
- Molecular Weight: 820.966761998
- Molecular Formula: C51H53N2O6P
- Purity: ≥98%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29321900
- Country Of Origin: China
- IUPAC: (2R,3S,5S)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(pyren-1-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@H]1C[C@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)C5=C6C7=C(C=C5)C=CC8=CC=CC(C=C6)=C78)OCCC#N)C(C)C)C

Catalog No. 50-235-5499 Supplier STA Pharmaceutical Supplier No. STAAM10140200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DNA α-pyrene amidite is a Amidite reagent (Subcategory: Specialty DNA Amidite) sold by WuXi TIDES. Offered in 200 mg. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 181227-52-1
- MDL: No data
- InChIKey: YYLVWADLYIVQFL-DGAGUGGYSA-N
- Molecular Weight: 820.966761998
- Molecular Formula: C51H53N2O6P
- Purity: ≥98%
- Container Type: 15 mL HDPE
- Pack Size: 200 mg
- Net Weight: 0.2 g
- Gross Weight: 6.9 g
- Commodity Code: 29321900
- Country Of Origin: China
- IUPAC: (2R,3S,5S)-2-((bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(pyren-1-yl)tetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: CC(N(P(O[C@H]1C[C@H](O[C@@H]1COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)C5=C6C7=C(C=C5)C=CC8=CC=CC(C=C6)=C78)OCCC#N)C(C)C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.