Learn More
Andwin Scientific DNA AWAY 12 EA/CSDNA AWAY 12
SDP
Catalog No. NC9888333
Encompass_Preferred
The MBP DNA AWAY removes DNase and DNA from laboratory surfaces. This is an efficient product for decontaminating apparatus, benchtops, glassware, and plasticware as it does not leave any residue to obstruct with gel polymerization or staining. It is packaged in a 250 mL squeeze bottle and sold in a case of 12.
This item is not returnable. View return policy
Catalog No. NC9888333 Supplier Andwin Scientific Supplier No. 53509506CS
The MBP DNA AWAY removes DNase and DNA from laboratory surfaces. This is an efficient product for decontaminating apparatus, benchtops, glassware, and plasticware as it does not leave any residue to obstruct with gel polymerization or staining. It is packaged in a 250 mL squeeze bottle and sold in a case of 12.