Learn More
Andwin Scientific DNA AWAY/ 250ML BOTTLE
SDP
Catalog No. NC0507591
Encompass_Preferred
The MBP DNA AWAY™ removes DNase and DNA from laboratory surfaces. This is an efficient product for decontaminating apparatus, benchtops, glassware, and plasticware as it does not leave any residue to obstruct with gel polymerization or staining. It is packaged and sold individually in a 250 mL squeeze bottle.
This item is not returnable. View return policy
Catalog No. NC0507591 Supplier Andwin Scientific Supplier No. 53509506250ML
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
The MBP DNA AWAY™ removes DNase and DNA from laboratory surfaces. This is an efficient product for decontaminating apparatus, benchtops, glassware, and plasticware as it does not leave any residue to obstruct with gel polymerization or staining. It is packaged and sold individually in a 250 mL squeeze bottle.