Learn More
Andwin Scientific DNA AWAY MBP 250ML
SDP
Catalog No. NC9785579
Encompass_Preferred
The MBP DNA AWAY removes DNase and DNA from laboratory surfaces. This is an efficient product for decontaminating apparatus, benchtops, glassware, and plasticware as it does not leave any residue to obstruct with gel polymerization or staining. It is packaged and sold individually in a 250 mL squeeze bottle.
Catalog No. NC9785579 Supplier Andwin Scientific Supplier No. E30711250ML
The MBP DNA AWAY removes DNase and DNA from laboratory surfaces. This is an efficient product for decontaminating apparatus, benchtops, glassware, and plasticware as it does not leave any residue to obstruct with gel polymerization or staining. It is packaged and sold individually in a 250 mL squeeze bottle.