Learn More
Cayman Chemical DNA Base ExcIsIonPath 10mg
SDP
Catalog No. 502972642
Encompass_Preferred
A cell-permeable inhibitor of Ape-1 (IC50 = 3-11 µM)
Catalog No. 50-297-2642 Supplier Cayman Chemical Supplier No. 1634810 MG
A cell-permeable inhibitor of Ape-1 (IC50 = 3-11 µM)