Learn More
Cayman Chemical DNA Base ExcIsIonPath 25mg
SDP
Catalog No. 502972431
Encompass_Preferred
A cell-permeable inhibitor of Ape-1 (IC50 = 3-11 µM)
Catalog No. 50-297-2431 Supplier Cayman Chemical Supplier No. 1634825 MG
A cell-permeable inhibitor of Ape-1 (IC50 = 3-11 µM)